Order Entry
Sweden
ContactUsLinkComponent
Anti-CTNNB1 Rabbit Polyclonal Antibody (CF568) [clone: Rabbit PAb]
Anti-CTNNB1 Rabbit Polyclonal Antibody (CF568) [clone: Rabbit PAb]
Leverantör:  Biotium
Certifikat
Anti-CTNNB1 Rabbit Polyclonal Antibody (CF568) [clone: Rabbit PAb]
Leverantör:  Biotium

Specifikationer

  • Catalog No:
  • BNC680376-100
  • BNC680376-500
  • Antigen symbol:
  • CTNNB1
  • CTNNB1
  • Antigen name:
  • Beta Catenin
  • Beta Catenin
  • Antigen synonyms:
  • CHBCAT|CATNB|beta 1 88kDa|Catenin beta-1|Cadherin associated protein)|CTNNB1
  • CHBCAT|CATNB|beta 1 88kDa|Catenin beta-1|Cadherin associated protein)|CTNNB1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • CF® 568
  • CF® 568
  • Clone:
  • Clone Rabbit PAb
  • Clone Rabbit PAb
  • Reactivity:
  • Human, Mouse
  • Human, Mouse
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 476018
  • 476018
  • Isotype:
  • IgG
  • IgG
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifikationer