Order Entry
Sweden
ContactUsLinkComponent
 
Anti-NKX2-2 Mouse Monoclonal Antibody (CF405S) [clone: NX2/294]
 :  Biotium
 
Anti-NKX2-2 Mouse Monoclonal Antibody (CF405S) [clone: NX2/294]
 :  Biotium

 

  • Catalog No:
  • BNC040294-100
  • BNC040294-500
  • Antigen symbol:
  • NKX2-2
  • NKX2-2
  • Antigen name:
  • NK2 homeobox 2
  • NK2 homeobox 2
  • Antigen synonyms:
  • NK2 transcription factor like protein B|NKX22|NK2 transcription factor related locus 2|Nkx2b|tinman|Homeobox protein NK-2 homolog B
  • NK2 transcription factor like protein B|NKX22|NK2 transcription factor related locus 2|Nkx2b|tinman|Homeobox protein NK-2 homolog B
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 405S
  • CF® 405S
  • Clone:
  • Clone NX2/294
  • Clone NX2/294
  • Reactivity:
  • Chicken, Human, Rat, Mouse
  • Chicken, Human, Rat, Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 516922
  • 516922
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide