Order Entry
Sweden
ContactUsLinkComponent
Anti-MAGEA1 Mouse Monoclonal Antibody (CF488A) [clone: MA454]
Anti-MAGEA1 Mouse Monoclonal Antibody (CF488A) [clone: MA454]
Supplier:  Biotium
Certificates
Anti-MAGEA1 Mouse Monoclonal Antibody (CF488A) [clone: MA454]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC880008-500
  • BNC880008-100
  • Antigen symbol:
  • MAGEA1
  • MAGEA1
  • Antigen name:
  • MAGE A1
  • MAGE A1
  • Antigen synonyms:
  • Melanoma associated antigen 1|CT1.1|Melanoma antigen family A 1|MAGE1A|MAGEA1|cancer/testis antigen 1.1|Melanoma associated antigen MZ2 E|MZ2 E
  • Melanoma associated antigen 1|CT1.1|Melanoma antigen family A 1|MAGE1A|MAGEA1|cancer/testis antigen 1.1|Melanoma associated antigen MZ2 E|MZ2 E
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 488A
  • CF® 488A
  • Clone:
  • Clone MA454
  • Clone MA454
  • Reactivity:
  • Human, Dog, Rat
  • Human, Dog, Rat
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 72879
  • 72879
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications