Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
D(-)-N-Methylglucamine
D-(-)-N-Methylglucamine, white powder
D-(-)-N-Methylglucamine, white powder
D-(-)-N-Methylglucamine, white powder
Leverantör:
MP Biomedicals
Certifikat
Expand image
D-(-)-N-Methylglucamine, white powder
Leverantör:
MP Biomedicals
Om denna produkt
CAS-nummer:
6284-40-8
:
Formel:
H₃CNHCH₂(CH(OH))₄CH₂OH
MDL-nummer:
MFCD00004707
:
195.22 g/mol
Smält Pt:
125…135 °C
Förvaringstemperatur:
Ambient