Aluminium sulphate hydrate, white powder ACS
:17927-65-0
:MFCD00149138
:Al₂(SO₄)₃·xH₂O
:MFCD00149136
:366.21 g/mol
:1.69 g/mL (25 °C) (lit.)
:Ambient
:233-135-0
:3077
:9,III
:GHS05
- Synonyms:Alum; Aluminum Trisulphate; Cake Alum
- Form:White Powder
- Storage:Store at Room Temperature (15 - 30 °C)
- Assay:98,0 - 102,0%
- Heavy Metals:≤0,001%
- Insoluble Matter:≤0,01%
- Chloride:≤0,005%
- Calcium:≤0,01%
- Magnesium:≤0,002%
- Potassium:≤0,005%
- Sodium:≤0,02%
- Iron (Fe):≤0,002%