Order Entry
Sweden
ContactUsLinkComponent
Ionomycin calcium salt
Ionomycin calcium salt
Supplier:  MP Biomedicals
Certificates
Ionomycin calcium salt
Supplier:  MP Biomedicals

Specification Test Results

  • Form:
    White powder
  • Storage:
    Store at +4 °C. Protect from light
  • Source:
    Streptomyces conglobatus

Specification Test Results