Order Entry
Sweden
ContactUsLinkComponent
2-Diethylaminoethanol ≥98%, Reagent Grade
2-Diethylaminoethanol ≥98%, Reagent Grade
Catalog # SPCMD1016-200LTBL
Supplier:  Spectrum Chemical
CAS Number:  
2-Diethylaminoethanol ≥98%, Reagent Grade
Catalog # SPCMD1016-200LTBL
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pk:
    200 L
  • Seq:
    5549

Specifications

Specification Test Results

  • Assay:
    Min. 98%
  • Storage Information:
    Hygroscopic: Keep tightly closed

Specification Test Results

About this item

 :
 :117.19 g/mol
UN:2686
ADR:8,II
 :GHS05
 :GHS02
CAS Number:100-37-8
Formula:C₆H₁₅NO
MDL Number:MFCD00002850