Order Entry
Sweden
Orders LinkContactUsLinkComponent
 

(±)-1,2-Propanediol, liquid

Leverantör:   MP Biomedicals
(±)-1,2-Propanediol, liquid
Leverantör:  MP Biomedicals
Certificates
(±)-1,2-Propanediol, liquid

2 Alternativ tillgängliga nedan

Product Details & Documents

Propylene glycol is a clear, colorless and hygroscopic liquid. Propylene glycol contains an asymmetrical carbon atom, so it exists in two stereoisomers.

The freezing point of water is depressed when mixed with propylene glycol due to increased opportunity for hydrogen bonding.
Propylene glycol is a solvent in many pharmaceuticals, including oral, injectable and topical formulations, such as for diazepam and lorazepam that are insoluble in water, use propylene glycol as a solvent in their clinical, injectable forms. It is also used as automotive antifreeze.

Synonymer:  Propylene glycol, 1,2-Propanediol, (±)-1,2-Propylene glycol, 1,2-Dihydroxypropane, (±)-1,2-Dihydroxy propane
  • CAS-nummer:57-55-6
  • Formel:CH₃CH(OH)CH₂OH
  • MDL-nummer:MFCD00064272
  • Molecular Weight:76.1 g/mol
  • Kokande Pt:187.9 °C (1013 hPa) (lit.)
  • Smält Pt:–60 °C (lit.)
  • Densitet:1.04 g/cm³ (20 °C) (lit.)
  • Förvaringstemperatur:Ambient
Specifications for Product Family

Specifikationstestresultat

  • Identity Test:
    Passes
  • FTIR:
    Conforms to Standard
Product Family Options

Produktinformation

  • Pk
    Tillgänglighet
    Price
  • 500 mL
    Log in för att kolla tillgänglighet och ert pris
    Log in för att kolla tillgänglighet och ert pris
    1 * 500 mL
    1 * 500 mL - 395,00 kr
    395,00 kr
    1 * 500 mL
       395,00 kr

  • 1 L
    Log in för att kolla tillgänglighet och ert pris
    Log in för att kolla tillgänglighet och ert pris
    1 * 1 L
    1 * 1 L - 580,00 kr
    580,00 kr
    1 * 1 L
       580,00 kr