Auramine O, dark yellow powder C.I. 41000
Catalog # ICNA0219506450
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pk:50 g
- Seq:3368
Specifications
Specification Test Results
- Identity Test :Passes
- UV/VIS: λmax (water):432±3 nm
Specification Test Results
About this item
:
:303.84 g/mol
UN:2811
ADR:6.1,III
CAS Number:2465-27-2
Formula:C₁₇H₂₂ClN₃
MDL Number:MFCD00012484