Order Entry
Sweden
ContactUsLinkComponent
Dextrin, white powder Commercial Grade, Type III
Dextrin, white powder Commercial Grade, Type III
Catalog # ICNA0216005790
Supplier:  MP Biomedicals
CAS Number:  
Dextrin, white powder Commercial Grade, Type III
Catalog # ICNA0216005790
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Identity Test:
    Passes
  • Source:
    Corn
  • Moisture Content:
    13% maximum
  • pH (12% aq. soln.):
    3.5 - 4.5

Specification Test Results

About this item

 :
 :180.16 g/mol
Storage Temperature:Ambient
CAS Number:9004-53-9
Formula:(C₆H₁₂O₆)n
MDL Number:MFCD00081554