Order Entry
Sweden
ContactUsLinkComponent
Sodium chloride ≥99.0%, white crystalline powder ACS
Sodium chloride ≥99.0%, white crystalline powder ACS
Supplier:  MP Biomedicals
Certificates
Sodium chloride ≥99.0%, white crystalline powder ACS
Supplier:  MP Biomedicals
Certificates

Specification Test Results

  • Assay:
    ≥99,0%
  • Barium:
    Passes
  • Bromide:
    ≤0,01%
  • Insoluble Matter:
    ≤0,005%
  • Magnesium (Mg):
    ≤0,001%
  • Sulphate (SO₄):
    ≤0,004%
  • Iron (Fe):
    ≤0,0002%
  • Calcium (Ca):
    ≤0,002%
  • Chlorates and Nitrates (as NO₃):
    ≤0,003%
  • Phosphate (PO₄):
    <0,0005%
  • Heavy Metals:
    ≤0,0005% (as Pb)
  • Potassium (K):
    ≤0,005%
  • Iodide (I):
    ≤0,002%
  • pH (5% aq soln, 25°C) :
    5.0 - 9.0

Specification Test Results

Related Information