Acrylamide ≥99%, crystalline powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:79-06-1
:
Formula:C₃H₅NO
MDL Number:MFCD00008032
:71.08 g/mol
UN:2074
ADR:6.1,III
:GHS06
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Identity Test:Passes
- Purity:≥99%
- FTIR:Conforms to Standard
- Melting Point:82 - 92 °C
- OD 290 nm (1% aq soln):≤0.10
- Acrylic acid content:≤0.01%