Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Urea
Urea ≥99%, white powder, Ultrapure
Urea ≥99%, white powder, Ultrapure
:
MP Biomedicals
Urea ≥99%, white powder, Ultrapure
:
MP Biomedicals
:
57-13-6
:
:
NH₂CONH₂
:
MFCD00008022
:
130…140 °C
:
60.06 g/mol