Skip to Header
Skip to Main Content
Skip to Footer
Avancerad
Avancerad
SDS
Certifikat
Tillverkardata
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Gum tragacanth
Gum tragacanth, powder
Gum tragacanth, powder
Gum tragacanth, powder
Supplier:
MP Biomedicals
Certificates
Expand image
Gum tragacanth, powder
Supplier:
MP Biomedicals
About this item
CAS Number:
9000-65-1
:
MDL Number:
MFCD00131255
Specification Test Results
Identity Test:
Passes
Specification Test Results