Whole wheat flour, powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:11006-34-1
:
Specification Test Results
- Form:Powder
- Storage:Store at Room Temperature (15 - 30 °C)
- Shelf Life:90 days at 70% relative humidity and good sanitary practices; 60 days best used