Saponin (from plant), white powder, Practical grade
Leverantör: MP Biomedicals
Certifikat
Om denna produkt
CAS-nummer:8047-15-2
:
MDL-nummer:MFCD00081981
:GHS07
Förvaringstemperatur:Omgivande
Specifikationstestresultat
- Identity Test:Passes
- Source:Plant
- FTIR:Conforms to Standard
- pH (5% aq soln):3-7
- Loss on Drying:≤10%