Order Entry
Sweden
ContactUsLinkComponent
Butyrolactone 3 ≥95%, crystalline solid
Butyrolactone 3 ≥95%, crystalline solid
Supplier:  Cayman Chemical
Certificates
Butyrolactone 3 ≥95%, crystalline solid
Supplier:  Cayman Chemical

Specification Test Results

  • Purity:
    ≥95%
  • Formal Name:
    rel-tetrahydro-4-methylene-5-oxo-2R-propyl-3S-furancarboxylic acid
  • Formulation:
    A Crystalline Solid
  • SMILES:
    O=C1C([C@@H](C(O)=O)[C@H](CCC)O1)=C
  • InChI Code:
    1S/C9H12O4/c1-3-4-6-7(8(10)11)5(2)9(12)13-6/h6-7H,2-4H2,1H3,(H,10,11)/t6-,7+/m0/s1
  • InChI Key:
    SRQUTZJZABSZRQ-NKWVEPMBSA-N
  • Storage:
    −20 °C
  • Shipping:
    Room temperature in continental US; may vary elsewhere
  • Stability:
    2 years from the QC date provided on the Certificate of Analysis, when stored properly

Specification Test Results