Order Entry
Sweden
ContactUsLinkComponent
 
D-(+)-Trehalose dihydrate 97.0-102.0% (by HPLC) NF, HPLE, multi-compendial parenteral grade, J.T.Baker®
 :  Avantor
 
undefined
D-(+)-Trehalose dihydrate 97.0-102.0% (by HPLC) NF, HPLE, multi-compendial parenteral grade, J.T.Baker®
 :  Avantor
 

 

  • Endotoxin Concentration, EU/g:
    ≤0.6
  • Trace Impurities - Aluminum (Al), For Information Only:
    = 0.6
  • Trace Impurities - Arsenic (As), For Information Only:
    For Information Only
  • Trace Impurities - Cadmium (Cd), For Information Only:
    For Information Only
  • Trace Impurities - Chromium (Cr), For Information Only:
    For Information Only
  • Trace Impurities - Copper (Cu), For Information Only:
    For Information Only
  • Trace Impurities - Iridium (Ir), For Information Only:
    For Information Only
  • Trace Impurities - Iron (Fe), For Information Only:
    For Information Only
  • Trace Impurities - Lead (Pb), For Information Only:
    For Information Only
  • Trace Impurities - Mercury (Hg), For Information Only:
    For Information Only
  • Trace Impurities - Manganese (Mn), For Information Only:
    For Information Only
  • Residual Methanol:
    ≤3000 ppm
  • Residual Ethanol:
    ≤5000 ppm
  • Residual Isopropanol:
    ≤5000 ppm
  • Cyanide (CN):
    ≤100 ppb
  • GMP Manufactured Product:
  • Meets BP. Chemical Specifications:
  • Meets EP Chemical Specifications:
  • Meets JP Chemical Specifications:
  • Meets NF Requirements:
  • CAUTION: For Manufacturing, Processing or Repackaging:
  • Bulk Pharmaceutical Chemical:
    97.0 - 102.0 %
  • NF - Assay (HPLC):
    97.0 - 102.0%
  • NF - Chloride (Cl):
    ≤0.0125%
  • NF - Colour and Clarity of Solution: Absorbance at 720 nm:
    ≤0.050
  • NF - Colour and Clarity of Solution, Absorption difference:
    ≤ 0.100
  • NF - Identification A:
    Passes Test
  • NF - Identification B:
    Passes Test
  • NF - Identification C:
    Passes Test
  • NF - Microbial Limits: TAMC (cfu/g):
    ≤100 cfu/g
  • NF - Microbial Limits: TYMC (cfu/g):
    ≤100 cfu/g
  • NF - Microbial Limits: E. coli:
    None Detected
  • NF - Microbial Limits: Salmonella:
    None Detected
  • NF - Nitrogen Content:
    ≤0.005%
  • NF - pH of 10% solution:
    4.5 - 6.5
  • NF - Related Substances - Total Peaks Eluting before Trehalose:
    ≤0.5%
  • NF - Related Substances - Total Peaks Eluting after Trehalose:
    ≤0.5%
  • NF - Residue on Ignition:
    ≤0.10%
  • NF - Soluble starch:
    Passes Test
  • NF - Specific Optical Rotation, Anhydrous α²ºD (+):
    197 - 201°
  • NF - Sulphate (SO₄):
    ≤0.0200%
  • NF - Water (by Karl Fischer Titration):
    9.0 - 11.0%
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - Assay (LC):
    97.0 - 102.0%
  • EP/BP - Chloride (Cl):
    ≤125 ppm
  • EP/BP - Identification A:
    Passes Test
  • EP/BP - Identification B:
    Passes Test
  • EP/BP - Identification C:
    Passes Test
  • EP/BP - Microbial Limits: TAMC (cfu/g):
    ≤100 cfu/g
  • EP/BP - Microbial Limits: TYMC (cfu/g):
    ≤100 cfu/g
  • EP/BP - Microbial Limits: E. coli:
    None Detected
  • EP/BP - Microbial Limits: Salmonella:
    None Detected
  • EP/BP - pH of 10% Solution:
    4.5 - 6.5
  • EP/BP - Related Substances - Impurity A:
    ≤0.5%
  • EP/BP - Related Substances - Impurity B:
    ≤ 0.5 %
  • EP/BP - Related Substances - Unspecified Impurities, each:
    ≤0.2%
  • EP/BP - Related Substances - Total Impurities:
    ≤1.0%
  • EP/BP - Soluble starch:
    Passes Test
  • EP/BP - Specific Optical Rotation, Anhydrous α²ºD (+):
    197 - 201°
  • EP/BP - Sulphate (SO₄):
    ≤200 ppm
  • EP/BP - Sulphated Ash:
    ≤0.1%
  • EP/BP - Water (by Karl Fischer Titration):
    9.0 - 11.0 %
  • JP - Assay (LC):
    98.0 - 101.0%
  • JP - Chloride (Cl):
    ≤0.018%
  • JP - Dextrin, Soluble Starch and Sulphite:
    Passes Test
  • JP - Heavy Metals (as Pb):
    ≤ 5 ppm
  • JP - Identification 1:
    Passes Test
  • JP - Identification 2:
    Passes Test
  • JP - Identification 3:
    Passes Test
  • JP - Nitrogen:
    ≤0.005%
  • JP - pH of 10% Solution:
    4.5 - 6.5
  • JP - Related Substances -Total Peaks Eluting before Trehalose:
    ≤0.5%
  • JP - Related Substances -Total Peaks Eluting after Trehalose:
    ≤0.5%
  • JP - Residue on Ignition:
    ≤0.1%
  • JP - Specific Optical Rotation, Anhydrous α²ºD (+):
    197 - 201°
  • JP - Sulphate (SO₄):
    ≤0.024%
  • JP - Water (by Karl Fischer Titration):
    9.0 - 11.0%
  • Appearance (White to Off White Powder):
    Passes Test
  • Conductivity, uS cm-¹:
    ≤15
  • Reducing Sugars:
    Passes Test
  • Glucose Area%:
    ≤0.5%
  • Microbial Testing - Staphylococcus aureus (USP):
    None Detected
  • Microbial Testing - Pseudomonas aeruginosa (USP):
    None Detected
  • Specific Optical Rotation, Dihydrate α²ºD (+):
    177.5 - 180.5°

 

Frequently Bought Together