Order Entry
Sweden
ContactUsLinkComponent
Melatonin, off-white powder
Melatonin, off-white powder
Leverantör:  MP Biomedicals
Certifikat
Melatonin, off-white powder
Leverantör:  MP Biomedicals
Specifikationstestresultat
  • Identity Test:
    Passes
  • Melting Point:
    114 - 124 °C
  • FTIR:
    Conforms to Standard
  • UV/Vis: λmax (methanol):
    222 - 225, 276 - 279 nm
  • Moisture:
    ≤1%
Specifikationstestresultat