Imidazole ≥99%, white crystalline powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:288-32-4
:
Formula:C₃H₄N₂
MDL Number:MFCD00005183
Boiling Pt:268 °C (lit.)
Melting Pt:86…94 °C
Density:500…600 kg/m³ (bulk density)
UN:2923
ADR:8,III
:68.08 g/mol
:GHS06
:GHS07
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Purity:≥99%
- FTIR:Conforms to Standard
- Melting Point:86 - 94 °C
- pH (1% aq.):≥7