Replica tapes, Press O-Film
Supplier: Industrial Physics
Certificates
About this item
This replica tape is specifically designed to measure blasted surfaces. By placing the Tqc replica tape on the surface and rubbing over it, the Rt (total roughness) or peak-to-valley height of the profile can be taken and measured.
Certifications: ASTM D4417-03