Anti-VNN1 Rabbit Polyclonal Antibody
Catalog # BSBTPB10106
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Vanin 1
- Antigen symbol:VNN1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:8876
- Antigen synonyms:Tiff66|HDLCQ8
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Pantetheinase(VNN1) detection. Tested with WB in Human.
Type: Primary
Antigen: VNN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human