Anti-NR2F6 Rabbit Polyclonal Antibody
Catalog # BSBTPB10081
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Nuclear Receptor Subfamily 2, Group F, Member 6
- Antigen symbol:NR2F6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Cross adsorption:No
- Gene ID:2063
- Antigen synonyms:EAR2|EAR-2|ERBAL2
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Nuclear receptor subfamily 2 group F member 6(NR2F6) detection. Tested with WB in Human;Rat.
Type: Primary
Antigen: NR2F6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat