Order Entry
Sweden
ContactUsLinkComponent
Anti-AKR1C1/C2 Rabbit Polyclonal Antibody
Anti-AKR1C1/C2 Rabbit Polyclonal Antibody
Catalog # BSBTPB10036
Supplier:  Boster Bio
undefined
Anti-AKR1C1/C2 Rabbit Polyclonal Antibody
Catalog # BSBTPB10036
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    AKR1C1/C2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    1645/1646
  • Size:
    100 μg
  • Pk:
    100 µG

Specifications

About this item

Rabbit IgG polyclonal antibody for Aldo-keto reductase family 1 member C1/C2(AKR1C1/C2) detection. Tested with WB in Human;Rat.

Type: Primary
Antigen: AKR1C1/C2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat