Anti-SPN39 Mouse Monoclonal Antibody [clone: SPN/839]
ABNOMAB13433
: Abnova
- Antibody type:Primary
- Antigen symbol:SPN39
- Clonality:Monoclonal
- Clone:SPN/839
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Epitope:Full-length
- Gene ID:6693
- Size:100 μg
- Pk:100 µG
Mouse monoclonal antibody raised against full length recombinant human SPN.
Type: Primary
Antigen: SPN39
Clonality: Monoclonal
Clone: SPN/839
Conjugation:
Epitope: Full-length
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human