Order Entry
Sweden
ContactUsLinkComponent
Anti-CD68/824 Mouse Monoclonal Antibody [clone: LAMP4/824]
Anti-CD68/824 Mouse Monoclonal Antibody [clone: LAMP4/824]
Catalog # ABNOMAB13481
Supplier:  Abnova
Anti-CD68/824 Mouse Monoclonal Antibody [clone: LAMP4/824]
Catalog # ABNOMAB13481
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    CD68/824
  • Clonality:
    Monoclonal
  • Clone:
    LAMP4/824
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Epitope:
    Full-length
  • Gene ID:
    968
  • Size:
    100 μg
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal antibody raised against full length recombinant human CD68.

Type: Primary
Antigen: CD68/824
Clonality: Monoclonal
Clone: LAMP4/824
Conjugation:
Epitope: Full-length
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human