Anti-CD441097 Mouse Monoclonal Antibody [clone: HCAM/1097]
Catalog # ABNOMAB13473
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen symbol:CD441097
- Clonality:Monoclonal
- Clone:HCAM/1097
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Epitope:Full-length
- Gene ID:960
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Mouse monoclonal antibody raised against full length recombinant human CD44.
Type: Primary
Antigen: CD441097
Clonality: Monoclonal
Clone: HCAM/1097
Conjugation:
Epitope: Full-length
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human