Order Entry
Sweden
ContactUsLinkComponent
Anti-ARG11125 Mouse Monoclonal Antibody [clone: ARG1/1125]
Anti-ARG11125 Mouse Monoclonal Antibody [clone: ARG1/1125]
Catalog # ABNOMAB13324
Supplier:  Abnova
Anti-ARG11125 Mouse Monoclonal Antibody [clone: ARG1/1125]
Catalog # ABNOMAB13324
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    ARG11125
  • Clonality:
    Monoclonal
  • Clone:
    ARG1/1125
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG3 kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    383
  • Size:
    100 μg
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal antibody raised against partial recombinant human ARG1.

Type: Primary
Antigen: ARG11125
Clonality: Monoclonal
Clone: ARG1/1125
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG3, kappa
Reactivity: Human