Anti-ARG11125 Mouse Monoclonal Antibody [clone: ARG1/1125]
Catalog # ABNOMAB13323
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen symbol:ARG11125
- Clonality:Monoclonal
- Clone:ARG1/1125
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG3 kappa
- Reactivity:Human
- Western blot:Yes
- Gene ID:383
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Mouse monoclonal antibody raised against partial recombinant human ARG1.
Type: Primary
Antigen: ARG11125
Clonality: Monoclonal
Clone: ARG1/1125
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG3, kappa
Reactivity: Human