Anti-IDH11152 Mouse Monoclonal Antibody [clone: IDH1/1152]
Catalog # ABNOMAB13317
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen symbol:IDH11152
- Clonality:Monoclonal
- Clone:IDH1/1152
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Gene ID:3417
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Mouse monoclonal antibody raised against partial recombinant human IDH1.
Type: Primary
Antigen: IDH11152
Clonality: Monoclonal
Clone: IDH1/1152
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human