Anti-Hspg2 Rat Monoclonal Antibody [clone: A7L6]
Catalog # ABNOMAB13313
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen symbol:Hspg2
- Clonality:Monoclonal
- Clone:A7L6
- Host:Rat
- ImmunoChemistry:Yes
- Isotype:IgG2a kappa
- Reactivity:Human,Mouse
- Gene ID:15530
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Rat monoclonal antibody raised against native mouse Hspg2.
Type: Primary
Antigen: Hspg2
Clonality: Monoclonal
Clone: A7L6
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a, kappa
Reactivity: Human,Mouse