Order Entry
Sweden
ContactUsLinkComponent
Anti-MAGEA1838 Mouse Monoclonal Antibody [clone: MZ2E/838]
Anti-MAGEA1838 Mouse Monoclonal Antibody [clone: MZ2E/838]
  ABNOMAB13339
 :  Abnova
Anti-MAGEA1838 Mouse Monoclonal Antibody [clone: MZ2E/838]
  ABNOMAB13339
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen symbol:
    MAGEA1838
  • Clonality:
    Monoclonal
  • Clone:
    MZ2E/838
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Epitope:
    Full-length
  • Gene ID:
    4100
  • Size:
    100 μg
  • Pk:
    100 µG

 

 

Mouse monoclonal antibody raised against full length recombinant human MAGEA1.

Type: Primary
Antigen: MAGEA1838
Clonality: Monoclonal
Clone: MZ2E/838
Conjugation:
Epitope: Full-length
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human