Order Entry
Sweden
ContactUsLinkComponent
Fast DNA™ SPIN kit, MP Biomedicals
Fast DNA™ SPIN kit, MP Biomedicals
Catalog # ICNA116540600
Supplier:  MP Biomedicals
CAS Number:  
undefined
Fast DNA™ SPIN kit, MP Biomedicals
Catalog # ICNA116540600
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Total DNA
  • Sample type:
    Plant samples
    Yeast/Fungi
    Cultured cells
    Invertebrates
    Human/animal cells and tissues
  • Target molecule:
    DNA
  • Description:
    Fast DNA™ SPIN Kit, 100×2 ml preps
  • Description:
    Fast DNA™ SPIN Kit

Specifications

About this item

The Fast DNA® SPIN Kit is used with the FastPrep®-24 or FastPrep® FP120 instrument to lyse and subsequently isolate DNA from up to 200 mg of almost any sample in less than 30 minutes.

  • Isolation of genomic DNA from plants, animals, bacteria, yeast, algae, and fungi using a silica spin filter method
  • Rapid and reproducible sample lysis with the FastPrep®-24 or FastPrep® FP120 instrument
  • Lyse and isolate DNA in less than 30 minutes from a variety of sample types
  • Flexible format eliminates need for separate DNA isolation kits
  • No hazardous organic reagents required
  • SPIN filters are included to streamline silica handling

Lysing Matrix A and three different chaotropic buffers are provided for the optimal homogenisation of a wide variety of sample types (plant and animal tissues, cultured cells, bacteria, yeast, fungi, insects, etc). The released DNA is purified by a silica-based GeneClean® SPIN procedure, with supplied filter buckets and catch tubes to simplify liquid handling, and is suitable for PCR analysis and other downstream applications.

For research purposes only.

Delivery: Kit contains Lysing Matrix A, 100×2,0 ml tubes;1/4" Ceramic Spheres, 100 spheres; Binding Matrix, 66 ml; Concentrated SEWS-M, 12 ml; DES, 25 ml; CLS-VF, 90 ml; PPS, 25 ml; CLS-TC, 110 ml; CLS-Y, 110 ml; SPIN Modules, 100 each; and Catch Tubes, 100 each.