Order Entry
Sweden
ContactUsLinkComponent
Magnetic stirring bars
Magnetic stirring bars
Catalog # HACHLZE136
Supplier:  Hach
undefined
Magnetic stirring bars
Catalog # HACHLZE136
Supplier:  Hach
Specifications
  • Ø:
    6 mm
  • Length:
    20 mm
  • Pk:
    5
Specifications
About this item
PFTE coated magnetic stirring bars.