Order Entry
Sweden
ContactUsLinkComponent
 
Test kits, photometric, nickel acid baths, LCK cuvette tests
Catalog # HACHLCK237
Supplier:  Hach
CAS Number:  
undefined
Test kits, photometric, nickel acid baths, LCK cuvette tests
Catalog # HACHLCK237
Supplier:  Hach
CAS Number:  

Specifications

  • Chemical method:
    Intrinsic colour of electroplating baths
  • Pk:
    25 Tests
  • Description:
    Nickel
  • Type:
    LCK237
  • For unit:
    DR6000, DR3900
  • Range:
    5 - 120 g/l

Specifications

About this item

Cuvette tests for acidic nickel plating baths, nickel sulphate and nickel sulphamate baths applications.

  • Precise and reliable measurement values
  • Maximum safety for users, thanks to the closed system and low amounts of reagents
  • Convenient and error-free dosing of the reagents
  • Barcode label for automatic recognition in the photometer
  • Differentiation between tests and measuring ranges by means of colour-coding

Shelf life: 12 months from production date

Storage conditions: 15 to 25 °C