Order Entry
Sweden
ContactUsLinkComponent
Cadmium(II) bromide tetrahydrate 98%, pure
Irritant
Environment
Cadmium(II) bromide tetrahydrate 98%, pure
Certificates
undefined
Irritant
Environment
Cadmium(II) bromide tetrahydrate 98%, pure

Specification Test Results

  • Appearance:
    White Powder or crystals
  • Assay:
    ≥97.5 %

Specification Test Results

Related Information