Order Entry
Sweden
ContactUsLinkComponent
Calcium stearate (6.4 - 7.4% Ca), powder
Calcium stearate (6.4 - 7.4% Ca), powder
Certificates
undefined
Calcium stearate (6.4 - 7.4% Ca), powder

Specification Test Results

  • Appearance:
    White to off-white Powder
  • Titration Complexometric:
    6.4 to 7.5 % (Ca)
  • Loss on drying:
    ≤6 % (105°C, 3 hrs)
  • Ash:
    9 % to 10.5 %

Specification Test Results

Related Information