Order Entry
Sweden
ContactUsLinkComponent
Carbon disulphide 99.9% ACS
Carbon disulphide 99.9% ACS
Katalog # ACRO447035000
CAS-nummer:  
undefined
Carbon disulphide 99.9% ACS
Katalog # ACRO447035000
CAS-nummer:  

Specifikationer

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifikationer

Specifikationstestresultat

  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.05 %
  • GC:
    ≥99.9 %
  • Color scale:
    ≤10 APHA
  • Residue after evaporation:
    ≤0.002 %
  • Sulfur dioxide (SO2):
    ≤2.5 ppm
  • Hydrogen sulfide (H2S):
    ≤1.5 ppm

Specifikationstestresultat

Om denna produkt

 :
 :76.14 g/mol
Kokande Pt:46 °C (760 mmHg)
Smält Pt:–111 °C
Densitet:1.262 g/cm³
Flash Pt:–30 °C
FN:1131
ADR:3,I
 :GHS07
 :GHS08
 :GHS02
Förvaringstemperatur:Ambient
CAS-nummer:75-15-0
Formel:CS₂
MDL-nummer:MFCD00011321