Order Entry
Sweden
ContactUsLinkComponent
Piperidine-2,4-dione 97%
Piperidine-2,4-dione 97%
Certifikat
undefined
Piperidine-2,4-dione 97%

Specifikationstestresultat

  • Appearance:
    White to yellow to light brown Powder or solid
  • Infrared spectrum:
    Conforms
  • Melting point:
    98 to 104 °C
  • NMR (1H-NMR):
    Consistent with structure

Specifikationstestresultat

Relaterad information