Calcium sulphate dihydrate 98+%, extra pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10101-41-4
:
Formula:CaSO₄·2H₂O
MDL Number:MFCD00149625
:172.17 g/mol
Density:2,32 g/cm³ (20 °C)
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Powder
- Titration Complexometric:≥98.0 %
- Water:19 to 23 %