Order Entry
Sweden
ContactUsLinkComponent
Ethanol 70%, Technical Grade (not denatured)
Flammable
Irritant
Ethanol 70%, Technical Grade (not denatured)
Catalog # ACRO444280050
CAS Number:  
undefined
Flammable
Irritant
Ethanol 70%, Technical Grade (not denatured)
Catalog # ACRO444280050
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Plastic drum
  • Pk:
    5 L
  • Seq:
    5190

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Density:
    (20°C) 0.878 to 0.893 kg/l
  • Ethanol:
    67 to 73 % v/v (calculated)
  • GC:
    ≥99.7 % (ethanol)

Specification Test Results

About this item

 :
 :46.07 g/mol
Boiling Pt:85…90 °C (760 mmHg)
Melting Pt:–48 °C
Flash Pt:21 °C°C
UN:1170
ADR:3,II
 :GHS02
 :GHS07
Storage Temperature:Ambient
CAS Number:64-17-5
Formula:C₂H₅OH
MDL Number:MFCD00003568