0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
N-TIPS pyrrole-3-boronic acid pinacol ester
N-TIPS pyrrole-3-boronic acid pinacol ester 95%
N-TIPS pyrrole-3-boronic acid pinacol ester 95%
:
Thermo Fisher Scientific
N-TIPS pyrrole-3-boronic acid pinacol ester 95%
:
Thermo Fisher Scientific
:
365564-11-0
:
:
C₁₉H₃₆BNO₂Si
:
MFCD08063117
:
349.397 g/mol
:
70…72 °C
:
GHS07
:
Ambient
Appearance:
White to brown Powder or solid
Infrared spectrum:
Conforms
GC:
≥94.0 %
ACRO44102