Eriochrome Black T ACS indicator grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1787-61-7
:
Formula:C₂₀H₁₂N₃NaO₇S
MDL Number:MFCD00003935
ADR:9,III
:461.39 g/mol
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Brown to brown-black Powder
- Infrared spectrum:Conforms
- Suitability:(as complexom. indicator) Passes test
- Clarity of solution:(0.1% water) Pases test