4-(Chloromethyl)benzyl alcohol 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16473-35-1
:
Formula:C₈H₉ClO
MDL Number:MFCD03427011
:156.609 g/mol
Melting Pt:58…60 °C
Storage Temperature:Ambient
UN:3261
ADR:8,II
:GHS05
Specification Test Results
- Appearance:White to light yellow Crystalline powder or solid
- Infrared spectrum:Conforms
- Melting point:57 to 61 °C
- GC:≥96.0 %