n-Pentane 95%, Technical Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:109-66-0
:
Formula:H₃C(CH₂)₃CH₃
MDL Number:MFCD00009498
:72.15 g/mol
Boiling Pt:36 °C (760 mmHg)
Melting Pt:–130 °C
Density:0.626 g/cm³
Flash Pt:–49 °C
UN:1265
ADR:3,II
:GHS07
:GHS08
:GHS09
:GHS02
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3570 to 1.3590 (20°C, 589 nm)
- GC:≥94 %
- UV:at 265 nm A: ≤1.0
- Residue after evaporation:≤0.001 %