



[2-(1-Pyrrolidinylmethyl)phenyl]magnesium bromide 0.25 M in THF, AcroSeal®
Catalog # 431650500.
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle, AcroSeal®
- Pk:50 mL
- Seq:2370
Specifications
Specification Test Results
- Appearance:Clear brown Solution
- Titration Grignard:0.15 to 0.35 M
- GC:≥98.5 %
Specification Test Results
About this item
:
Density:0.919 g/cm³
Flash Pt:–17 °C
ADR:3,II
:264.449 g/mol
Storage Temperature:Ambient
UN:2924
:GHS07
:GHS05
:GHS02
:GHS08
CAS Number:480424-80-4
Formula:C₁₁H₁₄BrMgN
MDL Number:MFCD03840891