Order Entry
Sweden
ContactUsLinkComponent
2-Bromobenzylmagnesium bromide 0.25 M in diethyl ether, AcroSeal®
Irritant
Corrosive
Flammable
2-Bromobenzylmagnesium bromide 0.25 M in diethyl ether, AcroSeal®
Catalog # 431611000.
CAS Number:  
undefined
Irritant
Corrosive
Flammable
2-Bromobenzylmagnesium bromide 0.25 M in diethyl ether, AcroSeal®
Catalog # 431611000.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, AcroSeal®
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Clear yellow to brown Solution
  • Titration Grignard:
    0.15 to 0.35 M
  • Melting point:
    84 to 87 °C
  • GC:
    ≥97.5 %

Specification Test Results

About this item

 :
 :274.24 g/mol
Density:0.766 g/cm³
Flash Pt:–40 °C
Storage Temperature:Ambient
UN:2924
ADR:3,II
 :GHS07
 :GHS05
 :GHS02
CAS Number:56812-60-3
Formula:C₇H₆Br₂Mg
MDL Number:MFCD01319903