


Dimethyldichlorosilane 99+%, AcroSeal®
:75-78-5
:
:C₂H₆Cl₂Si
:MFCD00000491
:129.06 g/mol
:70 °C (760 mmHg)
:–76 °C
:1.06 g/cm³
:–9 °C
:1162
:3,II
:GHS02
:GHS05
:GHS06
:Refrigerator
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4030 to 1.4060 (20°C, 589 nm)
- GC:≥99.0 %