Order Entry
Sweden
ContactUsLinkComponent
 
Hydrogen peroxide 3% (w/w) in water stabilised
 
undefined
Hydrogen peroxide 3% (w/w) in water stabilised
 

 

  • Appearance:
    Clear colorless Solution
  • Stabilizer:
    Approx. 300 ppm
  • Titration with KMnO4:
    3 to 4 %
  • GC:
    ≥97.5 %

 

Frequently Bought Together