
5-Bromo-2-chloroanisole 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16817-43-9
:
Formula:C₇H₆BrClO
MDL Number:MFCD00078659
:221.48 g/mol
Boiling Pt:240…245 °C (748 mmHg)
Melting Pt:27…28 °C
Density:1.631 g/cm³
Flash Pt:113 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid or low melting solid
- Infrared spectrum:Conforms
- GC:≥97.5 %